Please login to your online account to display your discounted pricing

(1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific uk ltd  CC46613DA

 View more versions of this product

Catalog No. CC46613DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 187.24
Quantity 1g
Assay 97%
CAS Max % 100.0
CAS 876728-39-1
Assay Percent Range 97%
Molecular Formula C11H13N3
CAS Min % 96.5
Packaging Amber Glass Bottle
Chemical Name or Material (1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD08271943

Specifications

Chemical Name or Material (1-Methyl-3-phenyl-1H-pyrazol-5-yl)methylamine
CAS 876728-39-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H13N3
Formula Weight 187.24
MDL Number MFCD08271943
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents