missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-pentylimidazolium Bromide, TCI America™

$134.45 - $452.23

Chemical Identifiers

CAS 343851-31-0
Molecular Formula C9H17BrN2
Molecular Weight (g/mol) 233.153
MDL Number MFCD19382537
InChI Key PXFKRXXEFJDOMO-UHFFFAOYSA-M
Synonym 1-Methyl-3-pentyl-1H-imidazol-3-ium Bromide
PubChem CID 10537569
IUPAC Name 1-methyl-3-pentylimidazol-1-ium;bromide
SMILES CCCCCN1C=C[N+](=C1)C.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M303725G View Documents TCI America
M303725G
25g Each for $452.23
 
M30375G View Documents TCI America
M30375G
5g Each for $134.45
 
Specifications

Chemical Identifiers

343851-31-0
233.153
PXFKRXXEFJDOMO-UHFFFAOYSA-M
10537569
CCCCCN1C=C[N+](=C1)C.[Br-]
C9H17BrN2
MFCD19382537
1-Methyl-3-pentyl-1H-imidazol-3-ium Bromide
1-methyl-3-pentylimidazol-1-ium;bromide

Specifications

Undesignated
343851-31-0
MFCD19382537
PXFKRXXEFJDOMO-UHFFFAOYSA-M
1-methyl-3-pentylimidazol-1-ium;bromide
10537569
Clear Liquid at 20°C
1-Methyl-3-pentylimidazolium Bromide
25g
C9H17BrN2
1-Methyl-3-pentyl-1H-imidazol-3-ium Bromide
CCCCCN1C=C[N+](=C1)C.[Br-]
233.153
233.15
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No