missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-3-pentylimidazolium Bromide 98.0+%, TCI America™
Supplier: TCI America M303725G
Specifications
1-Methyl-3-pentylimidazolium Bromide | |
C9H17BrN2 | |
25 g | |
PXFKRXXEFJDOMO-UHFFFAOYSA-M | |
1-methyl-3-pentylimidazol-1-ium;bromide | |
10537569 | |
≥98.0% (HPLC) |
343851-31-0 | |
MFCD19382537 | |
1-Methyl-3-pentyl-1H-imidazol-3-ium Bromide | |
CCCCCN1C=C[N+](=C1)C.[Br-] | |
233.153 | |
233.15 | |
Liquid |
Chemical Identifiers
343851-31-0 | |
233.153 | |
PXFKRXXEFJDOMO-UHFFFAOYSA-M | |
10537569 | |
CCCCCN1C=C[N+](=C1)C.[Br-] |
C9H17BrN2 | |
MFCD19382537 | |
1-Methyl-3-pentyl-1H-imidazol-3-ium Bromide | |
1-methyl-3-pentylimidazol-1-ium;bromide |
Safety and Handling
TSCA : No