Please login to your online account to display your discounted pricing

1-Methyl-3-octylimidazolium chloride, 97%, Acros Organics


Quantity 50g

Manufacturer: thermo fisher scientific  356610500

 View more versions of this product

Catalog No. AC356610500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 50g
Chemical Name or Material 1-Methyl-3-octylimidazolium chloride
CAS 64697-40-1
Assay Percent Range 97%
Molecular Formula C12H23ClN2
Formula Weight 230.78g/mol

Specifications

Chemical Name or Material 1-Methyl-3-octylimidazolium chloride
Assay Percent Range 97%
Molecular Formula C12H23ClN2
Formula Weight 230.78g/mol

Ionic liquid