Please login to your online account to display your discounted pricing

1-Methyl-3-octylimidazolium chloride, 97%, Acros Organics

$59.95 - $59.95

Chemical Identifiers

Chemical Name or Material 1-Methyl-3-octylimidazolium chloride
Assay Percent Range 97%
Molecular Formula C12H23ClN2
Formula Weight 230.78
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC356610050

  SDS
thermo fisher scientific
AC356610050
5g Each for $59.95
Description & Specifications

Specifications

Formula Weight 230.78
Quantity 5g
CAS 64697-40-1
Chemical Name or Material 1-Methyl-3-octylimidazolium chloride
Assay Percent Range 97%
Molecular Formula C12H23ClN2

Ionic liquid