missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-nitro-1-nitrosoguanidine (wetted with ca. 50% Water) (unit weight on dry weight basis), TCI America™

Manufacturer:  TCI America M05275G

 View more versions of this product

Catalog No. M05275G



Specifications

Specifications

1-Methyl-3-nitro-1-nitrosoguanidine (wetted with ca. 50% Water) (unit weight on dry weight basis)
5g
C2H5N5O3
2926
VZUNGTLZRAYYDE-UHFFFAOYSA-N
1-methyl-2-nitro-1-nitrosoguanidine
9562060
147.09
≥95.0% (T)
Undesignated
70-25-7
MFCD00007034
MNNG
CN(C(=N[N+](=O)[O-])N)N=O
147.094
CHEBI:21759
Crystal-Powder at 20°C

Chemical Identifiers

70-25-7
147.094
VZUNGTLZRAYYDE-UHFFFAOYSA-N
9562060
1-methyl-2-nitro-1-nitrosoguanidine
C2H5N5O3
MFCD00007034
MNNG
CHEBI:21759
CN(C(=N[N+](=O)[O-])N)N=O
Safety and Handling

Safety and Handling

RTECSNumber : MF4200000

TSCA : Yes

Recommended Storage : Refrigerator

Documents