missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-nitro-1-nitrosoguanidine (wetted with ca. 50% Water) (unit weight on dry weight basis), TCI America™

$135.97 - $478.20

Chemical Identifiers

CAS 70-25-7
Molecular Formula C2H5N5O3
Molecular Weight (g/mol) 147.094
MDL Number MFCD00007034
InChI Key VZUNGTLZRAYYDE-UHFFFAOYSA-N
Synonym MNNG
PubChem CID 9562060
ChEBI CHEBI:21759
IUPAC Name 1-methyl-2-nitro-1-nitrosoguanidine
SMILES CN(C(=N[N+](=O)[O-])N)N=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M05275G View Documents TCI America
M05275G
5g Each for $135.97
 
M052725G View Documents TCI America
M052725G
25g Each for $478.20
 
Specifications

Chemical Identifiers

70-25-7
147.094
VZUNGTLZRAYYDE-UHFFFAOYSA-N
9562060
1-methyl-2-nitro-1-nitrosoguanidine
C2H5N5O3
MFCD00007034
MNNG
CHEBI:21759
CN(C(=N[N+](=O)[O-])N)N=O

Specifications

Undesignated
70-25-7
MFCD00007034
MNNG
CN(C(=N[N+](=O)[O-])N)N=O
147.094
CHEBI:21759
Crystal-Powder at 20°C
1-Methyl-3-nitro-1-nitrosoguanidine (wetted with ca. 50% Water) (unit weight on dry weight basis)
5g
C2H5N5O3
2926
VZUNGTLZRAYYDE-UHFFFAOYSA-N
1-methyl-2-nitro-1-nitrosoguanidine
9562060
147.09
≥95.0% (T)
Safety and Handling

Safety and Handling

RTECSNumber : MF4200000

TSCA : Yes

Recommended Storage : Refrigerator