Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-3-n-octylimidazolium trifluoromethanesulfonate, 97%

Chemical Identifiers

Chemical Name or Material 1-Methyl-3-n-octylimidazolium trifluoromethanesulfonate
CAS 403842-84-2
Formula Weight 344.39
MDL Number MFCD06411717
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5959903

  SDS
alfa aesar™
H5959903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5959906

  SDS
alfa aesar™
H5959906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 344.39
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Sensitivity Moisture Sensitive
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 97%
Color Colorless to pale yellow
CAS 403842-84-2
Chemical Name or Material 1-Methyl-3-n-octylimidazolium trifluoromethanesulfonate
Health Hazard 1 Warning
Physical Form Low Melting Solid
MDL Number MFCD06411717
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.