missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-3-n-octylimidazolium hexafluorophosphate, 95%

$90.99 - $351.61

Chemical Identifiers

CAS 304680-36-2
Molecular Formula C12H23F6N2P
Molecular Weight (g/mol) 340.294
MDL Number MFCD06798203
InChI Key GRCIJNHHTXBJAK-UHFFFAOYSA-N
Synonym 1-methyl-3-n-octylimidazolium hexafluorophosphate, 3-methyl-1-octylimidazolium hexafluorophosphate, 1-methyl-3-octylimidazolium hexafluorophosphate, 1-octyl-3-methylimidazolium hexafluorophosphate, 1-methyl-3-n-octylimidazoliumhexafluorophosphate, acmc-20aj2z, dsstox_cid_27928, dsstox_rid_82681, dsstox_gsid_47952, ksc222c5l
PubChem CID 2734243
IUPAC Name 1-methyl-3-octylimidazol-1-ium;hexafluorophosphate
SMILES CCCCCCCCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5999006 View Documents Alfa Aesar™
H5999006
5g Each for $90.99
 
AAH5999014 View Documents Alfa Aesar™
H5999014
25g Each for $351.61
 
Specifications

Chemical Identifiers

304680-36-2
340.294
GRCIJNHHTXBJAK-UHFFFAOYSA-N
2734243
CCCCCCCCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F
C12H23F6N2P
MFCD06798203
1-methyl-3-n-octylimidazolium hexafluorophosphate, 3-methyl-1-octylimidazolium hexafluorophosphate, 1-methyl-3-octylimidazolium hexafluorophosphate, 1-octyl-3-methylimidazolium hexafluorophosphate, 1-methyl-3-n-octylimidazoliumhexafluorophosphate, acmc-20aj2z, dsstox_cid_27928, dsstox_rid_82681, dsstox_gsid_47952, ksc222c5l
1-methyl-3-octylimidazol-1-ium;hexafluorophosphate

Specifications

5g
C12H23F6N2P
1-methyl-3-n-octylimidazolium hexafluorophosphate, 3-methyl-1-octylimidazolium hexafluorophosphate, 1-methyl-3-octylimidazolium hexafluorophosphate, 1-octyl-3-methylimidazolium hexafluorophosphate, 1-methyl-3-n-octylimidazoliumhexafluorophosphate, acmc-20aj2z, dsstox_cid_27928, dsstox_rid_82681, dsstox_gsid_47952, ksc222c5l
CCCCCCCCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F
340.294
340.29
1-Methyl-3-n-octylimidazolium hexafluorophosphate
304680-36-2
MFCD06798203
GRCIJNHHTXBJAK-UHFFFAOYSA-N
1-methyl-3-octylimidazol-1-ium;hexafluorophosphate
2734243
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning