Learn More
1-Methyl-3-n-octylimidazolium chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5953406
Description
Role of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsRole of 1-methyl-3-octylimidazolium chloride in the micellization behavior of amphiphilic drug amitriptyline hydrochloride.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
1-Methyl-3-n-octylimidazolium chloride | |
Yellow | |
C12H23ClN2 | |
MFCD03095432 | |
Moisture sensitive | |
Slightly soluble in water. | |
[Cl-].CCCCCCCCN1C=C[N+](C)=C1 | |
230.78 | |
CHEBI:61342 | |
97% |
64697-40-1 | |
1.01 g/mL | |
1.51 | |
5 g | |
1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j | |
OXFBEEDAZHXDHB-UHFFFAOYSA-M | |
1-methyl-3-octylimidazol-1-ium;chloride | |
2734223 | |
230.78 | |
Liquid |
Chemical Identifiers
64697-40-1 | |
230.78 | |
OXFBEEDAZHXDHB-UHFFFAOYSA-M | |
2734223 | |
1-methyl-3-octylimidazol-1-ium;chloride |
C12H23ClN2 | |
MFCD03095432 | |
1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j | |
CHEBI:61342 | |
[Cl-].CCCCCCCCN1C=C[N+](C)=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only