missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-n-octylimidazolium Bromide, TCI America™

$146.67 - $436.95

Chemical Identifiers

CAS 61545-99-1
Molecular Formula C12H23BrN2
Molecular Weight (g/mol) 275.234
InChI Key URVSXZLUUCVGQM-UHFFFAOYSA-M
PubChem CID 10849985
IUPAC Name 1-methyl-3-octylimidazol-1-ium;bromide
SMILES CCCCCCCCN1C=C[N+](=C1)C.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19045G View Documents TCI America
M19045G
5g Each for $146.67
 
M190425G View Documents TCI America
M190425G
25g Each for $436.95
 
Specifications

Chemical Identifiers

61545-99-1
275.234
10849985
CCCCCCCCN1C=C[N+](=C1)C.[Br-]
C12H23BrN2
URVSXZLUUCVGQM-UHFFFAOYSA-M
1-methyl-3-octylimidazol-1-ium;bromide

Specifications

Yellow
61545-99-1
URVSXZLUUCVGQM-UHFFFAOYSA-M
1-methyl-3-octylimidazol-1-ium;bromide
10849985
Clear Liquid at 20°C
1-Methyl-3-n-octylimidazolium Bromide
5g
C12H23BrN2
CCCCCCCCN1C=C[N+](=C1)C.[Br-]
275.234
275.23
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No