missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-3-indoleacetic acid, 98%, ACROS Organics™

$63.58 - $63.58

Chemical Identifiers

CAS 1912-48-7
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
InChI Key NAIPEFIYIQFVFC-UHFFFAOYSA-N
Synonym 1-methyl-1 h-indol-3-yl acetic acid, 1-methyl-1h-indole-3-acetic acid, 1-methyl-3-indoleacetic acid, 1-methyl-3-indoleaceticacid, 1-methyl-3-indolyl acetic acid, 1-methylindol-3-yl acetic acid, 1-methylindole-3-acetic acid, 1h-indole-3-acetic acid, 1-methyl, 2-1-methyl-1h-indol-3-yl acetic acid, 2-1-methylindol-3-yl acetic acid
PubChem CID 254166
IUPAC Name 2-(1-methylindol-3-yl)acetic acid
SMILES CN1C=C(C2=CC=CC=C21)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC439240010
SDS View Documents
Thermo Fisher Scientific
AC439240010
1g Glass bottle
Each for $63.58
 
AC439240050
SDS View Documents
Thermo Fisher Scientific
AC439240050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1912-48-7
189.21
1-methyl-1 h-indol-3-yl acetic acid, 1-methyl-1h-indole-3-acetic acid, 1-methyl-3-indoleacetic acid, 1-methyl-3-indoleaceticacid, 1-methyl-3-indolyl acetic acid, 1-methylindol-3-yl acetic acid, 1-methylindole-3-acetic acid, 1h-indole-3-acetic acid, 1-methyl, 2-1-methyl-1h-indol-3-yl acetic acid, 2-1-methylindol-3-yl acetic acid
2-(1-methylindol-3-yl)acetic acid
C11H11NO2
NAIPEFIYIQFVFC-UHFFFAOYSA-N
254166
CN1C=C(C2=CC=CC=C21)CC(=O)O

Specifications

Authentic
Tan to White
1g
97.5
97.5% min. (HPLC)
1-methyl-1 h-indol-3-yl acetic acid, 1-methyl-1h-indole-3-acetic acid, 1-methyl-3-indoleacetic acid, 1-methyl-3-indoleaceticacid, 1-methyl-3-indolyl acetic acid, 1-methylindol-3-yl acetic acid, 1-methylindole-3-acetic acid, 1h-indole-3-acetic acid, 1-methyl, 2-1-methyl-1h-indol-3-yl acetic acid, 2-1-methylindol-3-yl acetic acid
CN1C=C(C2=CC=CC=C21)CC(=O)O
189.21
189.21
98%
Glass bottle
125.0°C to 126.5°C
1912-48-7
100.0
C11H11NO2
NAIPEFIYIQFVFC-UHFFFAOYSA-N
2-(1-methylindol-3-yl)acetic acid
254166
Crystalline Powder
1-Methyl-3-indoleacetic acid
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: rinse mouth.
Do

GHS Signal Word: Danger

SDS