missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-3-indoleacetic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC439240050
Specifications
1-Methyl-3-indoleacetic acid | |
Glass bottle | |
125.0°C to 126.5°C | |
1912-48-7 | |
100.0 | |
C11H11NO2 | |
NAIPEFIYIQFVFC-UHFFFAOYSA-N | |
2-(1-methylindol-3-yl)acetic acid | |
254166 | |
Crystalline Powder |
Authentic | |
Tan to White | |
5g | |
97.5 | |
97.5% min. (HPLC) | |
1-methyl-1 h-indol-3-yl acetic acid, 1-methyl-1h-indole-3-acetic acid, 1-methyl-3-indoleacetic acid, 1-methyl-3-indoleaceticacid, 1-methyl-3-indolyl acetic acid, 1-methylindol-3-yl acetic acid, 1-methylindole-3-acetic acid, 1h-indole-3-acetic acid, 1-methyl, 2-1-methyl-1h-indol-3-yl acetic acid, 2-1-methylindol-3-yl acetic acid | |
CN1C=C(C2=CC=CC=C21)CC(=O)O | |
189.21 | |
189.21 | |
98% |
Chemical Identifiers
1912-48-7 | |
189.21 | |
1-methyl-1 h-indol-3-yl acetic acid, 1-methyl-1h-indole-3-acetic acid, 1-methyl-3-indoleacetic acid, 1-methyl-3-indoleaceticacid, 1-methyl-3-indolyl acetic acid, 1-methylindol-3-yl acetic acid, 1-methylindole-3-acetic acid, 1h-indole-3-acetic acid, 1-methyl, 2-1-methyl-1h-indol-3-yl acetic acid, 2-1-methylindol-3-yl acetic acid | |
2-(1-methylindol-3-yl)acetic acid |
C11H11NO2 | |
NAIPEFIYIQFVFC-UHFFFAOYSA-N | |
254166 | |
CN1C=C(C2=CC=CC=C21)CC(=O)O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: rinse mouth.
Do
GHS Signal Word: Danger