Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-2-quinolinone, 99%

$69.27 - $227.81

Chemical Identifiers

Chemical Name or Material 1-Methyl-2-quinolinone
CAS 606-43-9
Molecular Formula C10H9NO
Formula Weight 159.19
MDL Number MFCD00024051
Beilstein 121318
Synonym 2-Hydroxy-1-methylquinoline; N-Methylcarbostyril
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0941004

  SDS
alfa aesar™
L0941004
2g Each for $69.27

AAL0941009

  SDS
alfa aesar™
L0941009
10g Each for $227.81
Description & Specifications

Specifications

Formula Weight 159.19
Chemical Name or Material 1-Methyl-2-quinolinone
CAS 606-43-9
Synonym 2-Hydroxy-1-methylquinoline; N-Methylcarbostyril
TSCA TSCA
Assay 99%
Molecular Formula C10H9NO
MDL Number MFCD00024051
Beilstein 121318
Melting Point 74° to 76°C
Quantity 2g