missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-2-pyrrolidone 99.0+%, TCI America™

Manufacturer:  Tci America M0418500ML

 View more versions of this product

Catalog No. M0418500ML



Specifications

Specifications

1-Methyl-2-pyrrolidone
Yellow
500mL
872-50-4
MFCD00003193
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
13387
99.13
≥99.0% (GC)
202°C
-24°C
7.7 - 8
C5H9NO
1-methyl-2-pyrrolidinone
CN1CCCC1=O
99.133
CHEBI:7307
Clear Liquid at 20°C

Chemical Identifiers

872-50-4
99.133
SECXISVLQFMRJM-UHFFFAOYSA-N
13387
1-methylpyrrolidin-2-one
C5H9NO
MFCD00003193
1-methyl-2-pyrrolidinone
CHEBI:7307
CN1CCCC1=O
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0113

RTECSNumber : UY5790000

TSCA : Yes

Documents