missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-2-Pyrrolidone, EMD Millipore

Manufacturer:  Millipore Sigma MX13901

 View more versions of this product

Catalog No. MMX13901



Specifications

Specifications

1-Methyl-2-Pyrrolidone
872-50-4
(CH3)-NC(:O)CH2CH2CH2-
SECXISVLQFMRJM-UHFFFAOYSA-N
1-methylpyrrolidin-2-one
13387
99.13
Glass Bottle
4L
OmniPure™
C5H9NO
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
CN1CCCC1=O
99.133
CHEBI:7307
Liquid
Undesignated

Chemical Identifiers

872-50-4
99.133
1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
CHEBI:7307
CN1CCCC1=O
C5H9NO
SECXISVLQFMRJM-UHFFFAOYSA-N
13387
1-methylpyrrolidin-2-one
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

Documents