Please login to your online account to display your discounted pricing

1-Methyl-2-pyrrolidone, 99.0% min., ACS, EMD Millipore

$668.60 - $668.60

Chemical Identifiers

Chemical Name or Material 1-Methyl-2-pyrrolidone
Name Note For ACS analysis
CAS 872-50-4
Purity Grade ACS Grade
Physical Form Liquid
Assay Percent Range 99.0% min.
Molecular Formula (CH3)-NC(:O)CH2CH2CH2
Linear Formula (CH3)-NC(:O)CH2CH2CH2-
Formula Weight 99.13
Synonym N-Methyl-2-pyrrolidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MMX13951

 
emd millipore corporation
MX13951
3kg Each for $668.60

MMX139530

 
emd millipore corporation
MX139530
203L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 99.13
Quantity 3kg
Color Clear
Physical Form Liquid
Name Note For ACS analysis
Assay Percent Range 99.0% min.
Molecular Formula (CH3)-NC(:O)CH2CH2CH2
Linear Formula (CH3)-NC(:O)CH2CH2CH2-
CAS 872-50-4
Recommended Storage Room Temperature
Chemical Name or Material 1-Methyl-2-pyrrolidone
Synonym N-Methyl-2-pyrrolidinone
Safety and Handling

Room Temperature