Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-2-pyrrolidinone-d9, 97% (Isotopic)

NMR Solvent

$689.59 - $689.59

Chemical Identifiers

Chemical Name or Material 1-Methyl-2-pyrrolidinone-d9
CAS 185964-60-7
Name Note Isotopic
Formula Weight 108.21
MDL Number MFCD00148961
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4233603

  SDS
alfa aesar™
4233603
1g Each for $689.59
Description & Specifications

Specifications

Formula Weight 108.21
Chemical Name or Material 1-Methyl-2-pyrrolidinone-d9
Name Note Isotopic
Boiling Point 81° to 82°C (10mmHg)
Melting Point −24°C
Physical Form Liquid
Assay 97%
Quantity 1g
Color Colorless
Flash Point 86°C (187°F)
MDL Number MFCD00148961
CAS 185964-60-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H315-H319-H335-H227
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P201-P309-P310
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P201-P309-P310
Obtain special instructions before use.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H360-H315-H319-H335-H227
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.