missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-2-pyrrolidinone, Biograde, 99.5%, Alfa Aesar™

Suitable for Biotech applications.Useful for spectrophotometry, chromatography and ICP-MS detection. Also, used for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide.

Manufacturer:  Thermo Fisher Scientific 44063K7

 View more versions of this product

Catalog No. AA44063K7



Description

Description

 Applications

  • Suitable for Biotech applications
  • Useful for spectrophotometry, chromatography and ICP-MS detection
  • Used for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide
  • Also useful for pharmaceutical, different industrial applications due to its broad solvency and low volatility
     

Specifications

Specifications

872-50-4
106420
99.13
Biochemical
270°C (518°F)
Under Argon
Miscible with water, lower alcohols, ketones, ethyl acetate, castor oil, chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
204°C
-24°C
Hygroscopic; Light sensitive
MFCD00003193
Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
Liquid,packaged under Argon
1.033 g/cm3 at 20°C
Amine-like
1.4705
86°C (187°F)
Colorless
4L

Chemical Identifiers

872-50-4
MFCD00003193
Safety and Handling

Safety and Handling

P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335-H360D-H373

EINECSNumber : 212-828-1

RTECSNumber : UY5790000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents