Learn More
1-Methyl-2-pyrrolidinone, ACS grade, 99.0+%, Thermo Scientific Chemicals
CAS Number-872-50-4, 4L, 106420, 204 deg.C, Colorless to Yellow, 1.033 g/mL, 212-828-1, 86 deg.C (187 deg.F), 99.13, ACS Reagent, H227-H315-H319-H335-H360D-H373, P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c, -24 deg.C
Supplier: Thermo Scientific Chemicals 043894AP
Description
1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Methyl-2-pyrrolidinone | |
872-50-4 | |
1.033 g/mL | |
500 mL | |
Amine-like | |
1.4705 | |
MFCD00003193 | |
Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons. | |
CN1CCCC1=O | |
99.13 | |
ACS Reagent |
-24°C | |
Colorless to Yellow | |
204°C | |
86°C (187°F) | |
Liquid | |
C5H9NO | |
106420 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
1-methylpyrrolidin-2-one | |
99.13 |
Chemical Identifiers
872-50-4 | |
99.13 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
CN1CCCC1=O |
C5H9NO | |
MFCD00003193 | |
1-methylpyrrolidin-2-one |
Safety and Handling
P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335-H360D-H373
EINECSNumber : 212-828-1
RTECSNumber : UY5790000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only