Please login to your online account to display your discounted pricing

1-Methyl-2-pyrrolidinone, 99.5%, for peptide synthesis, ACROS Organics™


Quantity 2.5L
Packaging Glass Bottle

Manufacturer: fisher scientific acros  AC354900025

 View more versions of this product

Catalog No. AC354900025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.4%
Physical Form Liquid
Formula Weight 99.13
Quantity 2.5L
Color Clear
Packaging Glass Bottle
Purity Grade For peptide synthesis
Density 1.0300g/mL
Fieser 01,696; 02,281; 09,316; 11,346
CAS Max % 100.0
Synonym N-Methylpyrrolidone
Free Amine 0.01% max.
MDL Number MFCD00003193
Assay Percent Range 99.5%
Molecular Formula C5H9NO
CAS 872-50-4
Chemical Name or Material 1-Methyl-2-pyrrolidinone
CAS Min % 99.4
Water 0.1% max. (K.F.)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Methyl-2-pyrrolidinone
CAS 872-50-4
CAS Min % 99.4
CAS Max % 100.0
Purity Grade For peptide synthesis
Assay ≥99.4%
Assay Percent Range 99.5%
Molecular Formula C5H9NO
Formula Weight 99.13
MDL Number MFCD00003193
Synonym N-Methylpyrrolidone
Accessories

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.

SDS & Safety Documents