Please login to your online account to display your discounted pricing

1-Methyl-2-pyrrolidinone, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™

$201.66 - $201.66

Chemical Identifiers

Chemical Name or Material 1-Methyl-2-pyrrolidinone
CAS 872-50-4
CAS Min % 99.4
CAS Max % 100.0
Purity Grade Extra Dry
Assay ≥99.4% (GC)
Assay Percent Range 99.5%
Molecular Formula C5H9NO
Formula Weight 99.13
MDL Number MFCD00003193
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC326930025

  SDS
fisher scientific acros
AC326930025
Glass Bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC326931000

  SDS
fisher scientific acros
326931000
Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC326930010

  SDS
fisher scientific acros
AC326930010
Glass Bottle 1L Each for $201.66
Description & Specifications

Specifications

Assay ≥99.4% (GC)
Physical Form Liquid
Formula Weight 99.13
Quantity 2.5L
Color Clear colorless
Packaging Glass Bottle
Water 0.005% Max. (Coulometric)
Purity Grade Extra Dry
Density 1.0300g/mL
Beilstein 21, 237
Fieser 01,696; 02,281; 09,316; 11,346
Merck Index 15, 6190
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99.5%
Molecular Formula C5H9NO
MDL Number MFCD00003193
CAS 872-50-4
Synonym N-Methylpyrrolidone
CAS Min % 99.4
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Filtered to 0.2 micrometer Yes, PFTE
Safety and Handling

Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May damage the unborn child.