Learn More
1-Methyl-2-pyrrolidinone, 98%, pure, ACROS Organics™
1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O
Manufacturer: Thermo Fisher Scientific AC449180050
Description
1-Methyl-2-pyrrolidinone, 98%, C5H9NO, CAS Number-872-50-4, 25L, 97.5% min. (GC), 202.0 deg.C, 1.0300g/mL, 212-828-1, 91 deg.C, 99.13, Pure, GHS Signal Word: Danger, SECXISVLQFMRJM-UHFFFAOYSA-N, 1-methylpyrrolidin-2-one, -24.0 deg.C, 15,619, Metal drum, 1.4690 to 1.4710, CN1CCCC1-O
Specifications
1-Methyl-2-pyrrolidinone | |
1.67 mPa.s (20°C) | |
97.5% min. (GC) | |
15,619 | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
1-methylpyrrolidin-2-one | |
99.13 | |
Pure | |
Metal drum | |
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents | |
202.0°C | |
5L |
1.03 | |
872-50-4 | |
C5H9NO | |
Solubility in water: 1000g/L (20°C). Other solubilities: miscible with most common organic solvents | |
CN1CCCC1=O | |
99.13 | |
98% | |
1.0300g/mL | |
1.4690 to 1.4710 | |
91°C | |
-24.0°C |
Chemical Identifiers
872-50-4 | |
99.13 | |
1-methylpyrrolidin-2-one |
C5H9NO | |
SECXISVLQFMRJM-UHFFFAOYSA-N | |
CN1CCCC1=O |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 212-828-1