Please login to your online account to display your discounted pricing

1-Methyl-2-pyrrolidineethanol 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC160870250

 View more versions of this product

Catalog No. AC160870250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay 97.5 to 102.5% Total Base, ≥97.5% (GC)
Physical Form Liquid
Formula Weight 129.2
Color Clear yellow to brownish
Quantity 25g
EINECS Number 266-538-5
TSCA TSCA
Density 0.95g/mL
Beilstein 21, I, 189
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4703 to 1.4723 (20°C, 589nm)
Water 0.5% max. (K.F.)
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-Methyl-2-pyrrolidineethanol
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H15NO
MDL Number MFCD00003174
CAS 67004-64-2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 1-Methyl-2-pyrrolidineethanol
Name Note 98%
CAS 67004-64-2
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5 to 102.5% Total Base, ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C7H15NO
Formula Weight 129.2
MDL Number MFCD00003174
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.

SDS & Safety Documents