missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-2-pyrrolidineethanol 98%, ACROS Organics™

$370.12 - $370.12

Chemical Identifiers

CAS 67004-64-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H15NO
Formula Weight 129.2
MDL Number MFCD00003174
Chemical Name or Material 1-Methyl-2-pyrrolidineethanol, 98%
Assay 97.5 to 102.5% Total Base, ≥97.5% (GC)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC160870250 SDS Thermo Fisher Scientific
AC160870250
Glass bottle 25g Each for $370.12
Specifications Description & Specifications

Chemical Identifiers

67004-64-2
100.0
C7H15NO
MFCD00003174
97.5 to 102.5% Total Base, ≥97.5% (GC)
97.5
98%
129.2
1-Methyl-2-pyrrolidineethanol, 98%

Specifications

25g
97.5
0.95g/mL
Glass bottle
Liquid
C7H15NO
129.2
21, I, 189
1-Methyl-2-pyrrolidineethanol, 98%
67004-64-2
100.0
Authentic
1.4703 to 1.4723 (20°C, 589nm)
98%
0.5% max. (K.F.)
MFCD00003174
Clear yellow to brownish
97.5 to 102.5% Total Base, ≥97.5% (GC)

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 266-538-5

TSCA : TSCA