missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-2-pyridone, TCI America™

Manufacturer:  TCI America M041225G

 View more versions of this product

Catalog No. M041225G



Specifications

Specifications

1-Methyl-2-pyridone
Yellow
694-85-9
MFCD00006520
DVVGIUUJYPYENY-UHFFFAOYSA-N
1-methylpyridin-2-one
12755
Clear Liquid at 20°C
252°C
25g
C6H7NO
1-methyl-2-pyridone, 1-methylpyridin-2 1h-one, n-methyl-2-pyridone, 2 1h-pyridinone, 1-methyl, 1-methyl-2 1h-pyridinone, n-methylpyridone, 1-methyl-2 1h-pyridone, 1-methyl-2-pyridinone, 2 1h-pyridone, 1-methyl, 1-methyl-1,2-dihydro-2-pyridinone
CN1C=CC=CC1=O
109.128
109.13
≥99.0% (GC)

Chemical Identifiers

694-85-9
109.128
DVVGIUUJYPYENY-UHFFFAOYSA-N
12755
CN1C=CC=CC1=O
C6H7NO
MFCD00006520
1-methyl-2-pyridone, 1-methylpyridin-2 1h-one, n-methyl-2-pyridone, 2 1h-pyridinone, 1-methyl, 1-methyl-2 1h-pyridinone, n-methylpyridone, 1-methyl-2 1h-pyridone, 1-methyl-2-pyridinone, 2 1h-pyridone, 1-methyl, 1-methyl-1,2-dihydro-2-pyridinone
1-methylpyridin-2-one
Safety and Handling

Safety and Handling

RTECSNumber : UV1148100

TSCA : Yes

Documents