missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-2-piperidinemethanol, 97%, ACROS Organics™

Manufacturer:  Acros Organics 168050250

 View more versions of this product

Catalog No. AC168050250



Specifications

Specifications

1-Methyl-2-piperidinemethanol
77°C
Glass bottle
79.0°C to 80.0°C (7.0mmHg)
25g
20845-34-5
100.0
C7H15NO
1-methyl-2-piperidinemethanol, 1-methylpiperidin-2-yl methanol, 1-methylpiperidine-2-methanol, 2-piperidinemethanol, 1-methyl, 1-methyl-2-piperidyl methanol, 1-methyl-2-hydroxymethylpiperidine, 2-hydroxymethyl-1-methylpiperidine, 1-methyl-2-piperidyl methan-1-ol, pubchem8873, 1-methyl-2piperidinemethanol
HXXJMMLIEYAFOZ-UHFFFAOYSA-N
(1-methylpiperidin-2-yl)methanol
89394
Liquid
0.9800g/mL
Authentic
1.4813 to 1.4833
Brown
97%
96.0
97%
MFCD00006494
Solubility in water: miscible. Other solubilities: soluble in most organic solvent
CN1CCCCC1CO
129.203
129.2
97%

Chemical Identifiers

20845-34-5
129.203
HXXJMMLIEYAFOZ-UHFFFAOYSA-N
89394
CN1CCCCC1CO
C7H15NO
MFCD00006494
1-methyl-2-piperidinemethanol, 1-methylpiperidin-2-yl methanol, 1-methylpiperidine-2-methanol, 2-piperidinemethanol, 1-methyl, 1-methyl-2-piperidyl methanol, 1-methyl-2-hydroxymethylpiperidine, 2-hydroxymethyl-1-methylpiperidine, 1-methyl-2-piperidyl methan-1-ol, pubchem8873, 1-methyl-2piperidinemethanol
(1-methylpiperidin-2-yl)methanol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 244-078-6

TSCA : TSCA

SDS Documents