Please login to your online account to display your discounted pricing

1-Methyl-2-piperidinemethanol, 97%, ACROS Organics™

$101.05 - $101.05

Chemical Identifiers

Chemical Name or Material 1-Methyl-2-piperidinemethanol
Name Note 97%
CAS 20845-34-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H15NO
Formula Weight 129.2
MDL Number MFCD00006494
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC168050250

  SDS
thermo fisher scientific
168050250
Glass bottle 25g Each for $101.05

AC168051000

  SDS
thermo fisher scientific
168051000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 129.2
Quantity 25g
Color Clear brown
EINECS Number 244-078-6
TSCA TSCA
Density 0.9800g/mL
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4813 to 1.4833 (20°C, 589nm)
Health Hazard 1 Danger
Assay Percent Range 97%
Molecular Formula C7H15NO
Name Note 97%
MDL Number MFCD00006494
Chemical Name or Material 1-Methyl-2-piperidinemethanol
CAS 20845-34-5
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.