missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-tetrazole, TCI America™

$125.28 - $392.64

Chemical Identifiers

CAS 16681-77-9
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.082
MDL Number MFCD00129972
InChI Key OMAFFHIGWTVZOH-UHFFFAOYSA-N
PubChem CID 140123
IUPAC Name 1-methyltetrazole
SMILES CN1C=NN=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24515G View Documents TCI America
M24515G
5g Each for $392.64
 
M24511G View Documents TCI America
M24511G
1g Each for $125.28
 
Specifications

Chemical Identifiers

16681-77-9
84.082
OMAFFHIGWTVZOH-UHFFFAOYSA-N
1-methyltetrazole
C2H4N4
MFCD00129972
140123
CN1C=NN=N1

Specifications

81°C
40°C
16681-77-9
MFCD00129972
OMAFFHIGWTVZOH-UHFFFAOYSA-N
1-methyltetrazole
140123
Crystal-Powder at 20°C
1-Methyl-1H-tetrazole
Undesignated
5g
C2H4N4
1325
CN1C=NN=N1
84.082
84.08
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No