Please login to your online account to display your discounted pricing

1-Methyl-1H-pyrrole-2-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-pyrrole-2-carboxylic acid
CAS 6973-60-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7NO2
Formula Weight 125.13
MDL Number MFCD00003088
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC02701DA

  SDS
fisher scientific company
CC02701DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC02701DE

 
fisher scientific company
CC02701DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 125.13
Quantity 1g
Assay 97%
CAS Max % 100.0
Molecular Formula C6H7NO2
MDL Number MFCD00003088
CAS 6973-60-0
Chemical Name or Material 1-Methyl-1H-pyrrole-2-carboxylic acid
Packaging Amber glass bottle
Assay Percent Range 97%
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.