missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-pyrazole-5-carboxylic acid, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC62301DA

 View more versions of this product

Catalog No. CC62301DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

126.11
97%
Amber Glass Bottle
16034-46-1
1-Methyl-1H-pyrazole-5-carboxylic acid
C5H6N2O2
1g
100.0
MFCD00464253
97%
96.5

Chemical Identifiers

16034-46-1
100.0
C5H6N2O2
MFCD00464253
97%
96.5
97%
126.11
1-Methyl-1H-pyrazole-5-carboxylic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning