Please login to your online account to display your discounted pricing

1-Methyl-1H-pyrazole-5-carbonyl chloride, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC62302CB

 View more versions of this product

Catalog No. CC62302CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 144.56
Quantity 250mg
Assay 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-5-carbonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H5ClN2O
CAS 84547-59-1
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 1-Methyl-1H-pyrazole-5-carbonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5ClN2O
Formula Weight 144.56
Accessories

GHS Signal Word: Danger