Please login to your online account to display your discounted pricing

1-Methyl-1H-pyrazole-5-carbonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-pyrazole-5-carbonyl chloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5ClN2O
Formula Weight 144.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC62302CB

  SDS
fisher scientific company
CC62302CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC62302DA

  SDS
fisher scientific company
CC62302DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.56
Quantity 250mg
Assay 97%
Chemical Name or Material 1-Methyl-1H-pyrazole-5-carbonyl chloride
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C5H5ClN2O
CAS 84547-59-1
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger