Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 98%

$39.18 - $39.18

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester
CAS 847818-74-0
Molecular Formula C10H17BN2O2
Formula Weight 208.07
MDL Number MFCD05861380
Synonym 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2709403

  SDS
alfa aesar™
H2709403
1g Each for $39.18
Description & Specifications

Specifications

Formula Weight 208.07
Synonym 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Quantity 1g
CAS 847818-74-0
Melting Point 58° to 61°C
MDL Number MFCD05861380
Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester
Assay 98%
Molecular Formula C10H17BN2O2

Applications
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester is used as a pharmaceutical intermediate, agrochemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.