Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester, 95%

$265.19 - $601.20

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester
CAS 1044851-76-4
Molecular Formula C9H15BN2O2
Formula Weight 194.04
MDL Number MFCD07777079
Synonym 5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2770303

  SDS
alfa aesar™
H2770303
1g Each for $601.20

AAH27703MD

  SDS
alfa aesar™
H27703MD
250mg Each for $265.19
Description & Specifications

Specifications

Formula Weight 194.04
Chemical Name or Material 1-Methyl-1H-pyrazole-5-boronic acid neopentyl glycol ester
Assay 95%
Molecular Formula C9H15BN2O2
MDL Number MFCD07777079
CAS 1044851-76-4
Synonym 5-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-pyrazole
Melting Point 72° to 75°C
Quantity 1g

Applications
Important intermediates and reagents in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.