missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%

Manufacturer:  Alfa Aesar™ H54785MD

 View more versions of this product

Catalog No. AAH54785MD



Specifications

Specifications

1-Methyl-1H-pyrazole-4-sulfonyl chloride
250mg
C4H5ClN2O2S
UN3265
RDAVKKQKMLINOH-UHFFFAOYSA-N
1-methylpyrazole-4-sulfonyl chloride
17024730
97%
Moisture sensitive
288148-34-5
MFCD04968667
1-methyl-1h-pyrazole-4-sulfonyl chloride
CN1C=C(C=N1)S(=O)(=O)Cl
180.606
179.98

Chemical Identifiers

288148-34-5
180.606
RDAVKKQKMLINOH-UHFFFAOYSA-N
17024730
CN1C=C(C=N1)S(=O)(=O)Cl
C4H5ClN2O2S
MFCD04968667
1-methyl-1h-pyrazole-4-sulfonyl chloride
1-methylpyrazole-4-sulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Store at -20°C

Documents