Learn More
1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5478503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Methyl-1H-pyrazole-4-sulfonyl chloride | |
C4H5ClN2O2S | |
1 g | |
Moisture sensitive | |
RDAVKKQKMLINOH-UHFFFAOYSA-N | |
1-methylpyrazole-4-sulfonyl chloride | |
17024730 | |
97% |
288148-34-5 | |
MFCD04968667 | |
UN3265 | |
1-methyl-1h-pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl, chloro 1-methylpyrazol-4-yl sulfone, 4-chlorosulphonyl-1-methyl-1h-pyrazole, 1-methylpyrazole-4-sulfonylchloride, 1-methyl-4-pyrazolesulfonyl chloride, 1h-pyrazole-4-sulfonylchloride, 1-methyl, 1h-pyrazole-4-sulfonylchloride,1-methyl-9ci, 4-chlorosulfonyl-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazole-4-sulphonyl chloride | |
CN1C=C(C=N1)S(=O)(=O)Cl | |
180.606 | |
179.98 |
Chemical Identifiers
288148-34-5 | |
180.606 | |
RDAVKKQKMLINOH-UHFFFAOYSA-N | |
17024730 | |
CN1C=C(C=N1)S(=O)(=O)Cl |
C4H5ClN2O2S | |
MFCD04968667 | |
1-methyl-1h-pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl, chloro 1-methylpyrazol-4-yl sulfone, 4-chlorosulphonyl-1-methyl-1h-pyrazole, 1-methylpyrazole-4-sulfonylchloride, 1-methyl-4-pyrazolesulfonyl chloride, 1h-pyrazole-4-sulfonylchloride, 1-methyl, 1h-pyrazole-4-sulfonylchloride,1-methyl-9ci, 4-chlorosulfonyl-1-methyl-1h-pyrazole, 1-methyl-1h-pyrazole-4-sulphonyl chloride | |
1-methylpyrazole-4-sulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only