missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Methyl-1H-pyrazole-4-boronic acid pinacol ester, 97%

Manufacturer:  Alfa Aesar™ H53139MD

 View more versions of this product

Catalog No. AAH53139MD



Description

Description

Applications
It is a reagent used for Suzuki-Miyaura cross-coupling reactions, transesterification reactions. It is involved in several reactions as a reagent for the preparation of aminothiazoles as γ-secretase modulators,amino-pyrido-indol-carboxamides, as potential JAK2 inhibitors for myeloproliferative disorders therapy, pyridine derivatives as TGF-β1 and active A signaling inhibitors and MK-2461 analogs as inhibitors of c-Met kinase for the treatment of cancer.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
Specifications

Specifications

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester
250mg
C10H17BN2O2
1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methylpyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methyl-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-4-pyrazoleboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C
208.068
208.07
59°C to 64°C
761446-44-0
MFCD03789259
UCNGGGYMLHAMJG-UHFFFAOYSA-N
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
2773987
97%

Chemical Identifiers

761446-44-0
208.068
UCNGGGYMLHAMJG-UHFFFAOYSA-N
2773987
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C
C10H17BN2O2
MFCD03789259
1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methylpyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-1h-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methyl-pyrazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-4-pyrazoleboronic acid pinacol ester
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents