missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Methyl-1H-pyrazole-4-boronic acid hydrochloride, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445490050
Specifications
1-Methyl-1H-pyrazole-4-boronic acid hydrochloride | |
1026796-02-0 | |
C4H7BN2O2·ClH | |
YFPLZINTZVVVJM-UHFFFAOYSA-N | |
(1-methylpyrazol-4-yl)boronic acid;hydrochloride | |
45158882 | |
95% |
5g | |
94% min. (HPLC) | |
1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride, 1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hcl, 1-methyl-1h-pyrazole-4-boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid xhcl, 1-methylpyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazole-4-boronic acid hcl, 1-methylpyrazole-4-boronic acid hydrochloride, 1-methylpyrazole-4-boronic acid, hcl, 1-methylpyrazole-4-boronic acid,hcl | |
B(C1=CN(N=C1)C)(O)O.Cl | |
162.38 | |
162.38 |
Chemical Identifiers
1026796-02-0 | |
162.38 | |
1-methyl-1h-pyrazol-4-yl boronic acid hydrochloride, 1-methyl-1h-pyrazol-4-ylboronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid hcl, 1-methyl-1h-pyrazole-4-boronic acid hydrochloride, 1-methyl-1h-pyrazole-4-boronic acid xhcl, 1-methylpyrazol-4-ylboronic acid hydrochloride, 1-methylpyrazole-4-boronic acid hcl, 1-methylpyrazole-4-boronic acid hydrochloride, 1-methylpyrazole-4-boronic acid, hcl, 1-methylpyrazole-4-boronic acid,hcl | |
(1-methylpyrazol-4-yl)boronic acid;hydrochloride |
C4H7BN2O2·ClH | |
YFPLZINTZVVVJM-UHFFFAOYSA-N | |
45158882 | |
B(C1=CN(N=C1)C)(O)O.Cl |
Safety and Handling
GHS Signal Word: Warning