Please login to your online account to display your discounted pricing

1-Methyl-1H-pyrazole-3-sulfonyl chloride, 97%, Maybridge

$545.15 - $545.15

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-pyrazole-3-sulfonyl chloride
CAS 89501-90-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H5ClN2O2S
Formula Weight 180.61
MDL Number MFCD08690273
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC48303DA

  SDS
fisher scientific company
CC48303DA
Amber Glass Bottle 1g Each for $545.15
Description & Specifications

Specifications

Formula Weight 180.61
Quantity 1g
Assay 97%
CAS Max % 100.0
MDL Number MFCD08690273
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Chemical Name or Material 1-Methyl-1H-pyrazole-3-sulfonyl chloride
CAS 89501-90-6
CAS Min % 96.5
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C4H5ClN2O2S
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.