missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-pyrazole-3-carboxylic acid, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC48301CB

 View more versions of this product

Catalog No. CC48301CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-Methyl-1H-pyrazole-3-carboxylic acid
94.5
95%
C5H6N2O2
250mg
YBFIKNNFQIBIQZ-UHFFFAOYSA-N
1-methylpyrazole-3-carboxylic acid
573176
126.11
25016-20-0
100.0
Amber Glass Bottle
MFCD00464254
1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid
CN1C=CC(=N1)C(O)=O
126.12
CHEBI:74738
95%

Chemical Identifiers

25016-20-0
126.12
YBFIKNNFQIBIQZ-UHFFFAOYSA-N
573176
1-methylpyrazole-3-carboxylic acid
C5H6N2O2
MFCD00464254
1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid
CHEBI:74738
CN1C=CC(=N1)C(O)=O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions