Please login to your online account to display your discounted pricing

(1-Methyl-1H-pyrazol-5-yl)methylamine, 97%, Maybridge

$180.04 - $180.04

Chemical Identifiers

Chemical Name or Material (1-Methyl-1H-pyrazol-5-yl)methylamine
CAS 863548-52-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H9N3
Formula Weight 111.15
MDL Number MFCD03419808
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

CC62313CB

 
fisher scientific company
CC62313CB
250mg Each for $180.04

CC62313DA

 
fisher scientific company
CC62313DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 111.15
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material (1-Methyl-1H-pyrazol-5-yl)methylamine
CAS 863548-52-1
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 97%
Molecular Formula C5H9N3
MDL Number MFCD03419808
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS P Statement Causes severe skin burns and eye damage.