missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1-Methyl-1H-pyrazol-3-yl)methylamine, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC48313CB

 View more versions of this product

Catalog No. CC48313CB

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

111.15
97%
612511-81-6
MFCD03419807
96.5
C5H9N3
250mg
100.0
95%
(1-Methyl-1H-pyrazol-3-yl)methylamine
Amber Glass Bottle

Chemical Identifiers

612511-81-6
100.0
C5H9N3
MFCD03419807
97%
96.5
95%
111.15
(1-Methyl-1H-pyrazol-3-yl)methylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger