Please login to your online account to display your discounted pricing

(1-Methyl-1H-pyrazol-3-yl)methylamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material (1-Methyl-1H-pyrazol-3-yl)methylamine
CAS 612511-81-6
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 95%
Molecular Formula C5H9N3
Formula Weight 111.15
MDL Number MFCD03419807
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC48313CB

  SDS
fisher scientific company
CC48313CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC48313DA

  SDS
fisher scientific company
CC48313DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 111.15
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material (1-Methyl-1H-pyrazol-3-yl)methylamine
Assay Percent Range 95%
MDL Number MFCD03419807
CAS 612511-81-6
Packaging Amber Glass Bottle
CAS Min % 96.5
Molecular Formula C5H9N3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.