Please login to your online account to display your discounted pricing

1-Methyl-1H-indole-7-carbonitrile, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC66916DA

 View more versions of this product

Catalog No. CC66916DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 156.19
Quantity 1g
Assay 97%
CAS Max % 100.0
CAS 52951-14-1
Chemical Name or Material 1-Methyl-1H-indole-7-carbonitrile
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C10H8N2
MDL Number MFCD09263864
CAS Min % 96.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Methyl-1H-indole-7-carbonitrile
CAS 52951-14-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H8N2
Formula Weight 156.19
MDL Number MFCD09263864
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.