missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-indole-7-carbonitrile, 97%, Maybridge

$242.35 - $242.35

Chemical Identifiers

CAS 52951-14-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H8N2
Formula Weight 156.19
MDL Number MFCD09263864
Chemical Name or Material 1-Methyl-1H-indole-7-carbonitrile
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC66916DA SDS Fisher Scientific Company
CC66916DA
Amber Glass Bottle 1g Each for $242.35
Specifications Description & Specifications

Chemical Identifiers

52951-14-1
100.0
C10H8N2
MFCD09263864
97%
96.5
97%
156.19
1-Methyl-1H-indole-7-carbonitrile

Specifications

52951-14-1
100.0
97%
156.19
1g
97%
96.5
Amber Glass Bottle
C10H8N2
MFCD09263864
1-Methyl-1H-indole-7-carbonitrile

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning