Please login to your online account to display your discounted pricing

1-Methyl-1H-indole-6-carbaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Methyl-1H-indole-6-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9NO
Formula Weight 159.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC44204CB

  SDS
fisher scientific company
CC44204CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC44204DA

  SDS
fisher scientific company
CC44204DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC44204DE

 
fisher scientific company
CC44204DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 159.19
Quantity 250mg
Assay 97%
Chemical Name or Material 1-Methyl-1H-indole-6-carbaldehyde
CAS 21005-45-8
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H9NO