missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-indole-5-sulfonyl chloride, 90%, Maybridge

$202.35 - $202.35

Chemical Identifiers

CAS 859850-75-2
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C9H8ClNO2S
Formula Weight 229.69
MDL Number MFCD08271907
Chemical Name or Material 1-Methyl-1H-indole-5-sulfonyl chloride
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC41403DA SDS Fisher Scientific Company
CC41403DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC41403CB SDS Fisher Scientific Company
CC41403CB
Amber Glass Bottle 250mg Each for $202.35
Specifications Description & Specifications

Chemical Identifiers

859850-75-2
100.0
C9H8ClNO2S
MFCD08271907
90%
89.5
90%
229.69
1-Methyl-1H-indole-5-sulfonyl chloride

Specifications

229.69
100.0
90%
C9H8ClNO2S
MFCD08271907
90%
1g
Amber Glass Bottle
859850-75-2
89.5
1-Methyl-1H-indole-5-sulfonyl chloride

Safety and Handling

GHS P Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh and keep confortable for several minutes.

Danger