Please login to your online account to display your discounted pricing

1-Methyl-1H-indole-5-sulfonyl chloride, 90%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC41403CB

 View more versions of this product

Catalog No. CC41403CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 229.69
Quantity 250mg
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh and keep confortable for several minutes.
Assay 90%
CAS Max % 100.0
Health Hazard 1 Danger
Chemical Name or Material 1-Methyl-1H-indole-5-sulfonyl chloride
Packaging Amber Glass Bottle
Molecular Formula C9H8ClNO2S
MDL Number MFCD08271907
CAS 859850-75-2
CAS Min % 89.5
Assay Percent Range 90%
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material 1-Methyl-1H-indole-5-sulfonyl chloride
CAS 859850-75-2
CAS Min % 89.5
CAS Max % 100.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C9H8ClNO2S
Formula Weight 229.69
MDL Number MFCD08271907
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh and keep confortable for several minutes.

GHS P Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.