missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Methyl-1H-indole-5-carboxylic acid, 95%, Maybridge

$92.77 - $92.77

Chemical Identifiers

CAS 186129-25-9
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD03839859
Chemical Name or Material 1-Methyl-1H-indole-5-carboxylic acid
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC41401CB SDS Fisher Scientific Company
CC41401CB
Amber Glass Bottle 250mg Each for $92.77
CC41401DA SDS Fisher Scientific Company
CC41401DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC41401DE Fisher Scientific Company
CC41401DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

186129-25-9
100.0
C10H9NO2
MFCD03839859
95%
94.5
95%
175.19
1-Methyl-1H-indole-5-carboxylic acid

Specifications

175.19
100.0
186129-25-9
MFCD03839859
95%
95%
250mg
Amber Glass Bottle
94.5
C10H9NO2
1-Methyl-1H-indole-5-carboxylic acid

Safety and Handling

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning